missing translation for 'onlineSavingsMsg'
Learn More

D-erythro-Sphingosine, 99+%

Catalog No. AAJ63584MA
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63584MA 10 mg
AAJ63584MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63584MA Supplier Thermo Scientific Chemicals Supplier No. J63584MA

CAS: 123-78-4 | C18H37NO2 | 299.499 g/mol

D-erythro-Sphingosine is used to inhibits protein kinase C and calmodulin-dependent enzymes, induces apoptosis. And also used to constituent of cell membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-erythro-Sphingosine is used to inhibits protein kinase C and calmodulin-dependent enzymes, induces apoptosis. And also used to constituent of cell membranes.

Solubility
Soluble in ethanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 123-78-4
Molecular Formula C18H37NO2
Molecular Weight (g/mol) 299.499
MDL Number MFCD00036751
InChI Key WWUZIQQURGPMPG-KRWOKUGFSA-N
Synonym sphingosine, d-erythro-sphingosine, 4-sphingenine, d-sphingosine, sphing-4-enine, cerebroside, sphingenine, 4-trans-sphingenine, trans-d-erythro-2-amino-4-octadecene-1,3-diol, 2s,3r,4e-2-amino-4-octadecene-1,3-diol
PubChem CID 5280335
ChEBI CHEBI:16393
IUPAC Name (E,2S,3R)-2-aminooctadec-4-ene-1,3-diol
SMILES CCCCCCCCCCCCCC=CC(C(CO)N)O

Specifications

Color White
Quantity 10 mg
Beilstein 1727294
Merck Index 14,8747
Solubility Information Soluble in ethanol.
Formula Weight 299.5
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material D-erythro-Sphingosine
EINECSNumber 204-651-3
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only