missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ D-erythro-Sphingosine, 99+%, synthetical
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99+% |
| Packaging | Glass bottle |
| Quantity | 50mg |
| Merck Index | 15, 8874 |
| Solubility Information | Solubility in water: insoluble in water. Other solubilities: insoluble in acetone, soluble in chloroform and dichloromethane |
| Formula Weight | 299.5 |
| Percent Purity | 99+% |
| Chemical Name or Material | D-erythro-Sphingosine, 99+% |
Safety and Handling
| EINECSNumber | 204-651-3 |
RUO – Research Use Only