missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Cysteine Hydrochloride Monohydrate 99.0+%, TCI America™
$208.78 - $605.83
Chemical Identifiers
| CAS | 207121-46-8 |
|---|---|
| Molecular Formula | C3H10ClNO3S |
| Molecular Weight (g/mol) | 175.627 |
| MDL Number | MFCD00150051 |
| InChI Key | QIJRTFXNRTXDIP-YBBRRFGFSA-N |
| Synonym | d-cysteine hydrochloride monohydrate, s-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, d-cysteine*hcl*h2o, d-cysteine hydrochloride hydrate, h-d-cys-oh.hcl.h2o, d-cysteine hydrate hydrochloride, c3h7no2s.hcl.h2o, d-cysteine hcl monohydrate, pubchem12314, h-d-cys-oh??hcl??h2o |
| PubChem CID | 10176341 |
| IUPAC Name | (2S)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride |
| SMILES | C(C(C(=O)O)N)S.O.Cl |
Chemical Identifiers
| 207121-46-8 | |
| 175.627 | |
| QIJRTFXNRTXDIP-YBBRRFGFSA-N | |
| 10176341 | |
| C(C(C(=O)O)N)S.O.Cl |
| C3H10ClNO3S | |
| MFCD00150051 | |
| d-cysteine hydrochloride monohydrate, s-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, d-cysteine*hcl*h2o, d-cysteine hydrochloride hydrate, h-d-cys-oh.hcl.h2o, d-cysteine hydrate hydrochloride, c3h7no2s.hcl.h2o, d-cysteine hcl monohydrate, pubchem12314, h-d-cys-oh??hcl??h2o | |
| (2S)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride |
Specifications
| 207121-46-8 | |
| C3H10ClNO3S | |
| 5 g | |
| QIJRTFXNRTXDIP-YBBRRFGFSA-N | |
| (2S)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride | |
| 10176341 | |
| ≥99.0% (T) | |
| D-Cysteine Hydrochloride Monohydrate |
| White | |
| MFCD00150051 | |
| d-cysteine hydrochloride monohydrate, s-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, d-cysteine*hcl*h2o, d-cysteine hydrochloride hydrate, h-d-cys-oh.hcl.h2o, d-cysteine hydrate hydrochloride, c3h7no2s.hcl.h2o, d-cysteine hcl monohydrate, pubchem12314, h-d-cys-oh??hcl??h2o | |
| C(C(C(=O)O)N)S.O.Cl | |
| 175.627 | |
| 157.61 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-3245
TSCA : No