missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Cysteine hydrochloride monohydrate, 98%
CAS: 207121-46-8 | C3H10ClNO3S | 175.627 g/mol
Supplier: Thermo Scientific Chemicals 442780010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| D-Cysteine hydrochloride monohydrate | |
| White to Yellow | |
| C3H10ClNO3S | |
| MFCD00150051 | |
| QIJRTFXNRTXDIP-YBBRRFGFSA-N | |
| (2S)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride | |
| 175.627 | |
| 177.77 |
| 207121-46-8 | |
| 98% | |
| Glass Bottle | |
| d-cysteine hydrochloride monohydrate, s-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, d-cysteine*hcl*h2o, d-cysteine hydrochloride hydrate, h-d-cys-oh.hcl.h2o, d-cysteine hydrate hydrochloride, c3h7no2s.hcl.h2o, d-cysteine hcl monohydrate, pubchem12314, h-d-cys-oh??hcl??h2o | |
| C(C(C(=O)O)N)S.O.Cl | |
| 1 g | |
| 10176341 | |
| Crystalline Powder |
Chemical Identifiers
| 207121-46-8 | |
| 175.627 | |
| QIJRTFXNRTXDIP-YBBRRFGFSA-N | |
| 10176341 | |
| C(C(C(=O)O)N)S.O.Cl |
| C3H10ClNO3S | |
| MFCD00150051 | |
| d-cysteine hydrochloride monohydrate, s-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, d-cysteine*hcl*h2o, d-cysteine hydrochloride hydrate, h-d-cys-oh.hcl.h2o, d-cysteine hydrate hydrochloride, c3h7no2s.hcl.h2o, d-cysteine hcl monohydrate, pubchem12314, h-d-cys-oh??hcl??h2o | |
| (2S)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride |
Safety and Handling
EINECSNumber : 251-043-9
RUO – Research Use Only