missing translation for 'onlineSavingsMsg'
Learn More

D-Cysteine, 98%

Catalog No. p-7054426
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6262703 1 g
AAH6262706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6262703 Supplier Thermo Scientific Chemicals Supplier No. H6262703

CAS: 921-01-7 | C3H7NO2S | 121.15 g/mol

D-Cysteine is an important source of sulfide in human metabolism. It is the precursor for glutathione. It plays important role in protein structure and in metal binding.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Cysteine is an important source of sulfide in human metabolism. It is the precursor for glutathione. It plays important role in protein structure and in metal binding.

Solubility
Soluble in water (partly).

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.

Chemical Identifiers

CAS 921-01-7
Molecular Formula C3H7NO2S
Molecular Weight (g/mol) 121.15
MDL Number MFCD00066461
InChI Key XUJNEKJLAYXESH-RUOZJJLTNA-N
Synonym d-cysteine, d-cystein, 2s-2-amino-3-sulfanylpropanoic acid, d-zystein, cysteine, d, s-2-amino-3-mercaptopropanoic acid, d-amino-3-mercaptopropionic acid, d-cys, h-d-cys-oh, cysteine
PubChem CID 92851
ChEBI CHEBI:16375
IUPAC Name (2S)-2-amino-3-sulfanylpropanoic acid
SMILES N[C@H](CS)C(O)=O

Specifications

Melting Point 230°C (decomposition)
Beilstein 1721407
Solubility Information Soluble in water (partly).
Quantity 1 g
Formula Weight 121.16
Percent Purity 98%
Sensitivity Air Sensitive
Physical Form Solid
Chemical Name or Material D-Cysteine
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P501c
EINECSNumber 213-062-0
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only