missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Cellotriose, 95%

Catalog No. AAJ67702LB0
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67702LB0 5 mg
AAJ67702MA 10 mg
AAJ67702MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ67702LB0 Supplier Thermo Scientific Chemicals Supplier No. J67702LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33404-34-1 | C18H32O16 | 504.44 g/mol

A 3-glucose cellulodextrin derived from cellulose degradation. An energy source for cellulosic bacteria. Can be used to help identify, differentiate and characterize oligosaccharide metabolizing enzymes such as endoglucanases and --glucosidases. Also useful in saccharification and ethanol fermentation research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33404-34-1
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.44
MDL Number MFCD00069842
InChI Key FYGDTMLNYKFZSV-ZWSAEMDYSA-N
Synonym cellotriose, d-+-cellotriose, polysaccharide, d-cellotriose, beta-d-glucopyranosyl-1->4-beta-d-glucopyranosyl-1->4-d-glucose, beta-d-glcp-1->4-beta-d-glcp-1->4-d-glc, beta-d-glucopyranosyl-1->4-beta-d-glucopyranosyl-1->4-d-glucopyranose, cellodextrin, d-glucose, o-d-glucopyranosyl, cellooligosaccharide
PubChem CID 5287993
ChEBI CHEBI:3528
IUPAC Name (2S,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@H]3[C@H](O)[C@@H](O)C(O)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Color White
Quantity 5 mg
Beilstein 100353
Sensitivity Hygroscopic
Solubility Information Soluble in water at 50mg/ml /n
Formula Weight 504.44
Percent Purity 95%
Physical Form Powder
Chemical Name or Material D-(+)-Cellotriose
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only