Learn More
Thermo Scientific Chemicals D-(+)-Cellotetraose, 95%
CAS: 38819-01-1 | C24H42O21 | 666.579 g/mol
Supplier: Thermo Scientific Chemicals J67583MB
Description
Derived from cellulose degradation. Used by cellulosic bacteria as an energy source. Useful for characterization and identification of oligosaccharide metabolizing enzymes such as endoglucanases and --glucosidases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Derived from cellulose degradation
- Used by cellulosic bacteria as an energy source
- Useful for characterization and identification of oligosaccharide metabolizing enzymes such as endoglucanases and _x001A_-glucosidases
Specifications
| D-(+)-Cellotetraose | |
| Plastic bottle | |
| MFCD00079079 | |
| 78869 | |
| cellotetraose | |
| LUEWUZLMQUOBSB-HQXNUCTASA-N | |
| (2S,3S,5S)-2-[(3S,5S,6S)-6-[(3S,5S,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(3S,5S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
| 131674892 | |
| 95% |
| 38819-01-1 | |
| C24H42O21 | |
| 25 mg | |
| Hygroscopic | |
| Soluble at 25mg/ml in water | |
| C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)O)CO)CO)CO)O)O)O)O | |
| 666.579 | |
| 666.58 | |
| Powder |
Chemical Identifiers
| 38819-01-1 | |
| 666.579 | |
| LUEWUZLMQUOBSB-HQXNUCTASA-N | |
| 131674892 | |
| C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)O)CO)CO)CO)O)O)O)O |
| C24H42O21 | |
| MFCD00079079 | |
| cellotetraose | |
| (2S,3S,5S)-2-[(3S,5S,6S)-6-[(3S,5S,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(3S,5S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only