missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Cellotetraose, 95%

Catalog No. aaj67583lb0
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67583LB0 5 mg
AAJ67583MA 10 mg
AAJ67583MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ67583LB0 Supplier Thermo Scientific Chemicals Supplier No. J67583LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 38819-01-1 | C24H42O21 | 666.579 g/mol

Derived from cellulose degradation. Used by cellulosic bacteria as an energy source. Useful for characterization and identification of oligosaccharide metabolizing enzymes such as endoglucanases and --glucosidases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Derived from cellulose degradation
  • Used by cellulosic bacteria as an energy source
  • Useful for characterization and identification of oligosaccharide metabolizing enzymes such as endoglucanases and _x001A_-glucosidases

Chemical Identifiers

CAS 38819-01-1
Molecular Formula C24H42O21
Molecular Weight (g/mol) 666.579
MDL Number MFCD00079079
InChI Key LUEWUZLMQUOBSB-HQXNUCTASA-N
Synonym cellotetraose
PubChem CID 131674892
IUPAC Name (2S,3S,5S)-2-[(3S,5S,6S)-6-[(3S,5S,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(3S,5S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)O)CO)CO)CO)O)O)O)O

Specifications

Color White
Quantity 5 mg
Beilstein 78869
Sensitivity Hygroscopic
Solubility Information Soluble at 25mg/mL in water
Formula Weight 666.58
Percent Purity 95%
Physical Form Powder
Chemical Name or Material D-(+)-Cellotetraose
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only