missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Cellobiose 98.0+%, TCI America™
$45.57 - $115.82
Chemical Identifiers
| CAS | 528-50-7 |
|---|---|
| Molecular Formula | C12H22O11 |
| Molecular Weight (g/mol) | 342.30 |
| MDL Number | MFCD00136034 |
| InChI Key | GUBGYTABKSRVRQ-UHFFFAOYNA-N |
| Synonym | d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose |
| PubChem CID | 57370084 |
| IUPAC Name | 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol |
| SMILES | OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O |
Chemical Identifiers
| C12H22O11 | |
| MFCD00136034 | |
| d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose | |
| 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol |
Specifications
| 528-50-7 | |
| C12H22O11 | |
| d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose | |
| OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O | |
| 342.30 | |
| 342.30 | |
| Crystalline Powder |
| White | |
| MFCD00136034 | |
| GUBGYTABKSRVRQ-UHFFFAOYNA-N | |
| 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol | |
| 57370084 | |
| ≥98.0% (HPLC) | |
| D-(+)-Cellobiose |
Safety and Handling
EINECSNumber : (5)-6960
TSCA : Yes