Learn More
United Scientific™ D Cell Battery Holder and Joiner
Description
- Pack of 4 D battery holders. Unique design allows units to be snapped together in parallel or series to extend capacity or voltage.
- D Battery Holders, Pack of 4
- Include male and female button snaps to allow series connections
- Include extensions on sides for parallel connections
Specifications
Specifications
| Product Type | Battery Holder |
| For Use With (Equipment) | D Batteries |
| For Use With (Application) | Electrical Experiments |
Warning: Cancer and reproductive harm