missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Calcium pantothenate, 98%

CAS: 137-08-6 | C18H32CaN2O10 | 476.54 g/mol

$71.36 - $406.12

Chemical Identifiers

CAS 137-08-6
Molecular Formula C18H32CaN2O10
Molecular Weight (g/mol) 476.54
MDL Number MFCD00002766
InChI Key FAPWYRCQGJNNSJ-DXHDTSSINA-L
Synonym calcium pantothenate, d-pantothenic acid hemicalcium salt, d-pantothenic acid, calcium salt, pantothenic acid, calcium salt, d, r-n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-beta-alanine calcium salt, beta-alanine, n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-, calcium salt, r
PubChem CID 131847364
IUPAC Name calcium;3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoic acid
SMILES [Ca++].CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O.CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC243300050
SDS View Documents
Thermo Scientific Chemicals
243300050
5 g Glass Bottle
Each for $71.36
Only null left
 
AC243301000
SDS View Documents
Thermo Scientific Chemicals
243301000
100 g Plastic Bottle
Each for $140.71
Only null left
 
AC243305000
SDS View Documents
Thermo Scientific Chemicals
243305000
500 g Plastic Bottle
Each for $406.12
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

137-08-6
476.54
FAPWYRCQGJNNSJ-DXHDTSSINA-L
131847364
[Ca++].CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O.CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O
C18H32CaN2O10
MFCD00002766
calcium pantothenate, d-pantothenic acid hemicalcium salt, d-pantothenic acid, calcium salt, pantothenic acid, calcium salt, d, r-n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-beta-alanine calcium salt, beta-alanine, n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-, calcium salt, r
calcium;3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoic acid

Specifications

190°C
White
5.0% max. (105°C, 3 hrs)
C18H32CaN2O10
[HOCH2C(CH3)2CHOHCONHCH2CH2COO]2Ca
+ 25
calcium pantothenate, d-pantothenic acid hemicalcium salt, d-pantothenic acid, calcium salt, pantothenic acid, calcium salt, d, r-n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-beta-alanine calcium salt, beta-alanine, n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-, calcium salt, r
FAPWYRCQGJNNSJ-DXHDTSSINA-L
calcium;3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoic acid
5 g
476.52
Powder
137-08-6
Authentic
8.2 to 8.6% (Ca) On dried substance (Complexometry)
Glass Bottle
MFCD00002766
15, 7118
Solubility in water: 350g/L (20°C). Other solubilities: soluble in glycerine,very slightly soluble in ethanol,insoluble in ether,chloroform
[Ca++].CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O.CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O
476.54
131847364
98%
D-Calcium pantothenate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-278-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only