missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Calcium pantothenate, 98%

Catalog No. AC243305000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC243305000 500 g Plastic Bottle
AC243300050 5 g Glass Bottle
AC243301000 100 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC243305000 Supplier Thermo Scientific Chemicals Supplier No. 243305000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 137-08-6 | C18H32CaN2O10 | 476.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 137-08-6
Molecular Formula C18H32CaN2O10
Molecular Weight (g/mol) 476.54
MDL Number MFCD00002766
InChI Key FAPWYRCQGJNNSJ-DXHDTSSINA-L
Synonym calcium pantothenate, d-pantothenic acid hemicalcium salt, d-pantothenic acid, calcium salt, pantothenic acid, calcium salt, d, r-n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-beta-alanine calcium salt, beta-alanine, n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-, calcium salt, r
PubChem CID 131847364
IUPAC Name calcium;3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoic acid
SMILES [Ca++].CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O.CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O

Specifications

Melting Point 190°C
Color White
Infrared Spectrum Authentic
Loss on Drying 5.0% max. (105°C, 3 hrs)
Assay Percent Range 8.2 to 8.6% (Ca) On dried substance (Complexometry)
Packaging Plastic Bottle
Linear Formula [HOCH2C(CH3)2CHOHCONHCH2CH2COO]2Ca
Specific Rotation + 25
Merck Index 15, 7118
Solubility Information Solubility in water: 350g/L (20°C). Other solubilities: soluble in glycerine,very slightly soluble in ethanol,insoluble in ether,chloroform
Specific Rotation Condition +25.00 (20.00°C c=5,H2O)
Quantity 500 g
Formula Weight 476.52
Percent Purity 98%
Physical Form Powder
Chemical Name or Material D-Calcium pantothenate
Show More Show Less
EINECSNumber 205-278-9

RUO – Research Use Only