missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D and C Yellow No. 10, Spectrum™ Chemical
Description
Spectrum™ Chemical D and C Yellow No. 10
Specifications
Specifications
| CAS Max % | 100% |
| Percent Purity | 85% |
| Grade | Reagent |
| Packaging | Amber Glass Bottle |
| Color | Yellow |
| Quantity | 25 g |
| Chemical Name or Material | D and C Yellow No. 10 |