missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D and C Red No. 33, Spectrum™ Chemical
Description
Spectrum™ Chemical D and C Red No. 33
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Amber Glass Bottle |
| Color | Red |
| Quantity | 5 kg |
| Grade | Reagent |
| Chemical Name or Material | D and C Red No. 33 |