missing translation for 'onlineSavingsMsg'
Learn More

D and C Orange No. 4, Spectrum™ Chemical

Catalog No. 18601973
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601973 100 g Amber Glass Bottle
18601974 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18601973 Supplier Spectrum Chemical Mfg Cor Supplier No. D1130100GM

D1130, 633-96-5, C16H11N2NaO4S

Spectrum™ Chemical D and C Orange No. 4

Chemical Identifiers

CAS 633-96-5
Molecular Formula C16H11N2NaO4S
Molecular Weight (g/mol) 350.32
InChI Key IGQFKZCLTLAWLO-UHFFFAOYSA-M
IUPAC Name sodium 4-[2-(2-oxo-1,2-dihydronaphthalen-1-ylidene)hydrazin-1-yl]benzene-1-sulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=C(NN=C2C(=O)C=CC3=CC=CC=C23)C=C1

Specifications

CAS Max % 100%
Grade Reagent
Packaging Amber Glass Bottle
Color Red
Quantity 100 g
Chemical Name or Material D and C Orange No. 4