missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Biotine, Fisher BioReagents
Description
La D-Biotine, qui forme un complexe stable avec l’Avidin, est utile pour l’étude des interactions Avidin-Biotine et en chromatographie d’affinité.
Spécifications
Spécifications
| Vecteur alpha | Biotine |
| Nom ou substance chimique | D-Biotin |
| Point de fusion | 228,9 °C |
| Couleur | Blanc |
| pH | 4.5 |
| Fourchette de pourcentages d’essai | ≥99 % |
| Emballage | Verre ambré |
| Quantité | 1 g |
| Indice de Merck | 15, 1236 |
| Rotation spécifique | +89 à +93° (+ ou -) |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. . NFPA Health:1 Flammability:0 Instability:0 |
| Recommended Storage | -2° à 8 °C au réfrigérateur |