missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Biotin, 98+%

Catalog No. AAA1420706
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1420706 5 g
AAA1420703 1 g
AAA1420709 10 g
AAA1420714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1420706 Supplier Thermo Scientific Chemicals Supplier No. A1420706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58-85-5 | C10H16N2O3S | 244.31 g/mol

Biotin is a form of vitamin B that helps the body to break down fats and carbohydrates. It is used as an alternative medicine to treat or prevent biotin deficiency, which is caused by malnutrition, rapid weight loss, long-term tube feeding and other medical conditions. It is an essential vitamin which is important for amino acid and energy metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in hot water, dimethyl sulfoxide, alcohol and benzene.

Notes
Store in cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 58-85-5
Molecular Formula C10H16N2O3S
Molecular Weight (g/mol) 244.31
MDL Number MFCD00005541
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
Synonym biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin
PubChem CID 171548
ChEBI CHEBI:15956
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12

Specifications

Melting Point 224°C to 226°C
Assay Percent Range ≥98%
Quantity 5 g
Beilstein 86838
Sensitivity Light sensitive
Merck Index 14,1231
Solubility Information Soluble in hot water,dimethyl sulfoxide,alcohol and benzene.
Formula Weight 244.32
Percent Purity ≥98%
Chemical Name or Material D-(+)-Biotin
EINECSNumber 200-399-3
RTECSNumber XJ9088200
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only