missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Biotin, ≥97.5%, Cell Culture Reagent Grade, MP Biomedicals™
Manufacturer: Mp Biomedicals 0219463401
Specifications
D-Biotin | |
1.267 g/cm3(Lit.) | |
≥97.5% | |
MFCD00005541 | |
1g | |
OC(=O)CCCCC1SCC2NC(=O)NC12 | |
244.31 | |
CHEBI:15956 | |
Crystalline Powder |
58-85-5 | |
White | |
C10H16N2O3S | |
biodermatin, bioepiderm, bios ii, biotin, coenzyme r, d +-biotin, d-biotin, factor s, vitamin b7, vitamin h | |
YBJHBAHKTGYVGT-UHFFFAOYNA-N | |
5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid | |
171548 | |
244.3 | |
Cell Culture Reagent |
Chemical Identifiers
58-85-5 | |
244.31 | |
YBJHBAHKTGYVGT-UHFFFAOYNA-N | |
171548 | |
5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid |
C10H16N2O3S | |
MFCD00005541 | |
biodermatin, bioepiderm, bios ii, biotin, coenzyme r, d +-biotin, d-biotin, factor s, vitamin b7, vitamin h | |
CHEBI:15956 | |
OC(=O)CCCCC1SCC2NC(=O)NC12 |
Safety and Handling
EINECSNumber : 200-399-3
Recommended Storage : Store at +4°C.