missing translation for 'onlineSavingsMsg'
Learn More

D-beta-Proline, 98+%

CAS: 72580-54-2 | C5H9NO2 | 115.132 g/mol

$172.84 - $519.00

Chemical Identifiers

CAS 72580-54-2
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD08272856
InChI Key JAEIBKXSIXOLOL-SCSAIBSYSA-N
Synonym r-pyrrolidine-3-carboxylic acid, 3r-pyrrolidine-3-carboxylic acid, r---pyrrolidine-3-carboxylic acid, d-beta-proline, r-beta-proline, 3-pyrrolidinecarboxylic acid, 3r, r-3-pyrrolidinecarboxylic acid, l-pyrrolidine-3-carboxylic acid, 3r-3-pyrrolidinecarboxylic acid hcl, r-pyrrilidine-3-carboxylic acid
PubChem CID 1501970
IUPAC Name (3R)-pyrrolidine-3-carboxylic acid
SMILES C1CNCC1C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH57356MD
View Documents
Thermo Scientific Chemicals
H57356MD
250 mg
Each for $172.84
Only null left
 
AAH5735603
View Documents
Thermo Scientific Chemicals
H5735603
1 g
Each for $519.00
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

72580-54-2
115.132
JAEIBKXSIXOLOL-SCSAIBSYSA-N
1501970
C1CNCC1C(=O)O
C5H9NO2
MFCD08272856
r-pyrrolidine-3-carboxylic acid, 3r-pyrrolidine-3-carboxylic acid, r---pyrrolidine-3-carboxylic acid, d-beta-proline, r-beta-proline, 3-pyrrolidinecarboxylic acid, 3r, r-3-pyrrolidinecarboxylic acid, l-pyrrolidine-3-carboxylic acid, 3r-3-pyrrolidinecarboxylic acid hcl, r-pyrrilidine-3-carboxylic acid
(3R)-pyrrolidine-3-carboxylic acid

Specifications

185°C to 191°C
C5H9NO2
5496144
JAEIBKXSIXOLOL-SCSAIBSYSA-N
(3R)-pyrrolidine-3-carboxylic acid
115.132
−18° (c=2 in Water)
≥98%
72580-54-2
MFCD08272856
r-pyrrolidine-3-carboxylic acid, 3r-pyrrolidine-3-carboxylic acid, r---pyrrolidine-3-carboxylic acid, d-beta-proline, r-beta-proline, 3-pyrrolidinecarboxylic acid, 3r, r-3-pyrrolidinecarboxylic acid, l-pyrrolidine-3-carboxylic acid, 3r-3-pyrrolidinecarboxylic acid hcl, r-pyrrilidine-3-carboxylic acid
C1CNCC1C(=O)O
250 mg
1501970
115.13
D-beta-Proline
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only