missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ D(-)-Aspartic acid, 99+%

Supplier:  Thermo Scientific™ 204260050

Catalog No. AC204260050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

D(-)-Aspartic acid
1783-96-6
0.4% max. (105°C, 3 hrs)
0.3% max. (TLC)
Glass bottle
MFCD00063081
− 25.80
15, 830
CKLJMWTZIZZHCS-UWTATZPHSA-N
(2R)-2-aminobutanedioic acid
133.103
83887
133.1
99+%
Authentic
99+%
C4H7NO4
HO2CCH2CH(NH2)CO2H
0.1% max.
04, 471
d-aspartic acid, r-2-aminosuccinic acid, d-aspartate, h-d-asp-oh, r-aspartic acid, --aspartic acid, aspartic acid d-form, 2r-2-aminobutanedioic acid, d---aspartic acid, aspartic acid, d
C(C(C(=O)O)N)C(=O)O
− 25.80 (20.00°C c=5,5N HCl)
5g
CHEBI:17364
≥99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 217-234-6

RTECSNumber : CI9097500

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only