missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(-)-Asparagine monohydrate, 99%

Catalog No. AAB2455614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2455614 25 g
AAB2455622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2455614 Supplier Thermo Scientific Chemicals Supplier No. B2455614

CAS: 5794-24-1 | C4H10N2O4 | 150.134 g/mol

D-(-)-Asparagine monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-Asparagine monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in water (35 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 5794-24-1
Molecular Formula C4H10N2O4
Molecular Weight (g/mol) 150.134
MDL Number MFCD00149558
InChI Key RBMGJIZCEWRQES-HSHFZTNMSA-N
Synonym d-asparagine monohydrate, d-asparagine hydrate, r-2,4-diamino-4-oxobutanoic acid hydrate, d-asparagine, monohydrate, d-asparaginemonohydrate, d---asparagine monohydrate, d-aspartic acid 4-amide, r---2-aminosuccinamic acid, d--asparagine monohydrate, r-2-aminosuccinic acid 4-amide
PubChem CID 16211197
IUPAC Name (2R)-2,4-diamino-4-oxobutanoic acid;hydrate
SMILES C(C(C(=O)O)N)C(=O)N.O

Specifications

Melting Point 274°C to 276°C (decomposition)
Merck Index 14,837
Solubility Information Soluble in water (35g/L at 20°C).
Quantity 25 g
Formula Weight 150.14 (132.12 Anhydrous)
Percent Purity 99%
Chemical Name or Material D-(-)-Asparagine monohydrate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only