missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Asparagine monohydrate
Supplier: Thermo Scientific Chemicals 204270010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 280.0°C | |
| 5794-24-1 | |
| H2NCOCH2CH(NH2)CO2H·H2O | |
| 14, 837 | |
| RBMGJIZCEWRQES-UHFFFAOYNA-N | |
| 2-amino-3-carbamoylpropanoic acid hydrate | |
| 150.13 |
| D-Asparagine monohydrate | |
| C4H10N2O4 | |
| 04, 471 | |
| Solubility aq. soln.: 35 g/L (20°C). Other solubilities: soluble in acids and alkalies, practically insoluble in methanol, ethanol, ether, and benzene | |
| O.NC(CC(N)=O)C(O)=O | |
| 1 g | |
| 150.13 |
Chemical Identifiers
| 5794-24-1 | |
| RBMGJIZCEWRQES-UHFFFAOYNA-N | |
| O.NC(CC(N)=O)C(O)=O |
| 150.13 | |
| 2-amino-3-carbamoylpropanoic acid hydrate |
RUO – Research Use Only