missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Asparagine monohydrate

Catalog No. AC204270250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC204270250 25 g
AC204270010 1 g
AC204270050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC204270250 Supplier Thermo Scientific Chemicals Supplier No. 204270250

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5794-24-1
Molecular Weight (g/mol) 150.13

Specifications

Melting Point 280.0°C
Molecular Formula C4H8N2O3·H2O
Linear Formula H2NCOCH2CH(NH2)CO2H·H2O
Beilstein 04, 471
Merck Index 14, 837
Solubility Information Solubility aq. soln.: 35 g/L (20°C). Other solubilities: soluble in acids and alkalies, practically insoluble in methanol, ethanol, ether, and benzene
Quantity 25 g
Formula Weight 150.13
Chemical Name or Material D-Asparagine monohydrate

RUO – Research Use Only