missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(-)-Arginine 98.0+%, TCI America™
$96.84 - $334.16
Chemical Identifiers
| CAS | 157-06-2 |
|---|---|
| Molecular Formula | C6H14N4O2 |
| Molecular Weight (g/mol) | 174.204 |
| MDL Number | MFCD00063116 |
| InChI Key | ODKSFYDXXFIFQN-SCSAIBSYSA-N |
| Synonym | d-arginine, h-d-arg-oh, r-2-amino-5-guanidinopentanoic acid, d-arginin, d-arg, d---arginine, arginine, d, d-2-amino-5-guanidinovaleric acid, d--arginine, unii-r54z304z7c |
| PubChem CID | 71070 |
| ChEBI | CHEBI:15816 |
| IUPAC Name | (2R)-2-amino-5-(diaminomethylideneamino)pentanoic acid |
| SMILES | C(CC(C(=O)O)N)CN=C(N)N |
Chemical Identifiers
| 157-06-2 | |
| 174.204 | |
| ODKSFYDXXFIFQN-SCSAIBSYSA-N | |
| 71070 | |
| (2R)-2-amino-5-(diaminomethylideneamino)pentanoic acid |
| C6H14N4O2 | |
| MFCD00063116 | |
| d-arginine, h-d-arg-oh, r-2-amino-5-guanidinopentanoic acid, d-arginin, d-arg, d---arginine, arginine, d, d-2-amino-5-guanidinovaleric acid, d--arginine, unii-r54z304z7c | |
| CHEBI:15816 | |
| C(CC(C(=O)O)N)CN=C(N)N |
Specifications
| 157-06-2 | |
| C6H14N4O2 | |
| d-arginine, h-d-arg-oh, r-2-amino-5-guanidinopentanoic acid, d-arginin, d-arg, d---arginine, arginine, d, d-2-amino-5-guanidinovaleric acid, d--arginine, unii-r54z304z7c | |
| C(CC(C(=O)O)N)CN=C(N)N | |
| 1 g | |
| 71070 | |
| 174.20 | |
| Crystalline Powder |
| White | |
| MFCD00063116 | |
| ODKSFYDXXFIFQN-SCSAIBSYSA-N | |
| (2R)-2-amino-5-(diaminomethylideneamino)pentanoic acid | |
| 174.204 | |
| CHEBI:15816 | |
| ≥98.0% (T) | |
| D-(-)-Arginine |
Safety and Handling
TSCA : Yes