missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Arginine 98.0+%, TCI America™

Catalog No. A10291G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10291G 1 g
A10295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10291G Supplier TCI America Supplier No. A10291G

Chemical Identifiers

CAS 157-06-2
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.204
MDL Number MFCD00063116
InChI Key ODKSFYDXXFIFQN-SCSAIBSYSA-N
Synonym d-arginine, h-d-arg-oh, r-2-amino-5-guanidinopentanoic acid, d-arginin, d-arg, d---arginine, arginine, d, d-2-amino-5-guanidinovaleric acid, d--arginine, unii-r54z304z7c
PubChem CID 71070
ChEBI CHEBI:15816
IUPAC Name (2R)-2-amino-5-(diaminomethylideneamino)pentanoic acid
SMILES C(CC(C(=O)O)N)CN=C(N)N

Specifications

Color White
Quantity 1 g
Formula Weight 174.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-(-)-Arginine