missing translation for 'onlineSavingsMsg'
Learn More

D-Arginine Methyl Ester Dihydrochloride 98.0+%, TCI America™

Catalog No. p-7117689
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20161G 1 g
A20165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20161G Supplier TCI America Supplier No. A20161G

Chemical Identifiers

CAS 78851-84-0
Molecular Formula C7H18Cl2N4O2
Molecular Weight (g/mol) 261.147
MDL Number MFCD00153417
InChI Key XQYZOBNLCUAXLF-ZJIMSODOSA-N
PubChem CID 12714066
IUPAC Name methyl (2R)-2-amino-5-(diaminomethylideneamino)pentanoate;dihydrochloride
SMILES COC(=O)C(CCCN=C(N)N)N.Cl.Cl

Specifications

Color White
Quantity 1 g
Synonym h-d-arg-ome.2hcl, h-d-arg-ome 2hcl, r-methyl 2-amino-5-guanidinopentanoate dihydrochloride, d-arginine methyl ester dihydrochloride, d-arginine methyl ester di hydrochloride, d-arginine, methyl ester, dihydrochloride, methyl 2r-2-amino-5-carbamimidamidopentanoate dihydrochloride, h-d-arg-ome 2 hcl, h-d-arg-ome. 2hcl, d-arginine methyl ester dihydrochlorid
Formula Weight 261.15
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material D-Arginine Methyl Ester Dihydrochloride