Learn More
Thermo Scientific Chemicals D-Arginine, 98%
CAS: 157-06-2 | C6H14N4O2 | 174.204 g/mol
$128.12 - $470.05
Chemical Identifiers
| CAS | 157-06-2 |
|---|---|
| Molecular Formula | C6H14N4O2 |
| Molecular Weight (g/mol) | 174.204 |
| MDL Number | MFCD00063116 |
| InChI Key | ODKSFYDXXFIFQN-SCSAIBSYSA-N |
| Synonym | d-arginine, h-d-arg-oh, r-2-amino-5-guanidinopentanoic acid, d-arginin, d-arg, d---arginine, arginine, d, d-2-amino-5-guanidinovaleric acid, d--arginine, unii-r54z304z7c |
| PubChem CID | 71070 |
| ChEBI | CHEBI:15816 |
| IUPAC Name | (2R)-2-amino-5-(diaminomethylideneamino)pentanoic acid |
| SMILES | C(CC(C(=O)O)N)CN=C(N)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1613703
|
Thermo Scientific Chemicals
A1613703 |
1 g |
Each for $128.12
|
|
|||||
|
AAA1613706
|
Thermo Scientific Chemicals
A1613706 |
5 g |
Each for $470.05
|
|
|||||
Description
1-Deamino-8-D-arginine vasopressin makes a new pharmacological approach to the management of haemophilia and von Willebrand's disease. A novel nonenzymatic pathway for the generation of nitric oxide is by the reaction of hydrogen peroxide and d- or l-arginine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Deamino-8-D-arginine vasopressin makes a new pharmacological approach to the management of haemophilia and von Willebrand’s disease. A novel nonenzymatic pathway for the generation of nitric oxide is by the reaction of hydrogen peroxide and d- or l-arginine.
Solubility
Insoluble in water.
Notes
Air Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
Chemical Identifiers
| 157-06-2 | |
| 174.204 | |
| ODKSFYDXXFIFQN-SCSAIBSYSA-N | |
| 71070 | |
| (2R)-2-amino-5-(diaminomethylideneamino)pentanoic acid |
| C6H14N4O2 | |
| MFCD00063116 | |
| d-arginine, h-d-arg-oh, r-2-amino-5-guanidinopentanoic acid, d-arginin, d-arg, d---arginine, arginine, d, d-2-amino-5-guanidinovaleric acid, d--arginine, unii-r54z304z7c | |
| CHEBI:15816 | |
| C(CC(C(=O)O)N)CN=C(N)N |
Specifications
| ∼235°C | |
| C6H14N4O2 | |
| 1725412 | |
| Soluble in water at 148g/L. | |
| C(CC(C(=O)O)N)CN=C(N)N | |
| 174.204 | |
| 71070 | |
| 174.2 | |
| Air Sensitive |
| 157-06-2 | |
| MFCD00063116 | |
| d-arginine, h-d-arg-oh, r-2-amino-5-guanidinopentanoic acid, d-arginin, d-arg, d---arginine, arginine, d, d-2-amino-5-guanidinovaleric acid, d--arginine, unii-r54z304z7c | |
| ODKSFYDXXFIFQN-SCSAIBSYSA-N | |
| (2R)-2-amino-5-(diaminomethylideneamino)pentanoic acid | |
| 1 g | |
| CHEBI:15816 | |
| 98% | |
| D-Arginine |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
EINECSNumber : 205-866-5
RTECSNumber : CF1934220
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only