missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Amygdalin hydrate, 95%

Catalog No. AC104730050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC104730050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC104730050 Supplier Thermo Scientific Chemicals Supplier No. 104730050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 29883-15-6 | C20H27NO11 | 457.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29883-15-6
Molecular Formula C20H27NO11
Molecular Weight (g/mol) 457.43
MDL Number MFCD29049822
InChI Key XUCIJNAGGSZNQT-ARGBPYSKNA-N
Synonym amygdalin
PubChem CID 66509032
IUPAC Name (2R)-2-phenyl-2-[(3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxyacetonitrile
SMILES OC[C@H]1O[C@@H](OC[C@H]2O[C@@H](OC(C#N)C3=CC=CC=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 223.0°C to 226.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 17, V,8, 118
Merck Index 15, 592
Specific Rotation − 38.50
Solubility Information (4% in water) Clear
Formula Weight 457.42
Percent Purity 95%
Physical Form Fine Crystalline Powder
Chemical Name or Material D(-)-Amygdalin hydrate, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 249-925-3

RUO – Research Use Only