missing translation for 'onlineSavingsMsg'
Learn More

D-Amygdalin, 98%

Catalog No. AAJ6147214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6147214 25 g
AAJ6147203 1 g
AAJ6147206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6147214 Supplier Thermo Scientific Chemicals Supplier No. J6147214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A benzylic glycoside that has been used as an antineoplastic agent | CAS: 29883-15-6 | C20H27NO11 | 457.43 g/mol

A benzylic glycoside that has been used as an antineoplastic agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29883-15-6
Molecular Formula C20H27NO11
Molecular Weight (g/mol) 457.43
MDL Number MFCD29049822
InChI Key XUCIJNAGGSZNQT-ARGBPYSKNA-N
Synonym amygdalin
PubChem CID 66509032
IUPAC Name 2-phenyl-2-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxan-2-yl]oxy}acetonitrile
SMILES OC[C@H]1O[C@@H](OC[C@H]2O[C@@H](OC(C#N)C3=CC=CC=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 210°C to 215°C
Color White
Quantity 25 g
Beilstein 66856
Merck Index 14,597
Optical Rotation −38° (c=1.2 in Water)
Formula Weight 457.44
Percent Purity 98%
Physical Form Powder
Chemical Name or Material D-Amygdalin
Hazard Category H302
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 249-925-3
RTECSNumber OO8450000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only