missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-alpha-Tocopherylquinone 97,0+%, TCI America™
Spécifications
Spécifications
| Quantity | 500 mg |
| Formula Weight | 446.72 |
| Percent Purity | ≥97.0% (HPLC) |
| Physical Form | Liquide |
| Chemical Name or Material | D-alpha-Tocopherylquinone |
Sécurité et manipulation
| RTECSNumber | DK5170000 |
| TSCA | No |
| Recommended Storage | Réfrigérateur |