missing translation for 'onlineSavingsMsg'
Learn More

D-alpha-Tocopherol Succinate 97.0+%, TCI America™

Catalog No. p-7137326
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26285G 5 g
T262825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26285G Supplier TCI America Supplier No. T26285G
Only null left

Chemical Identifiers

CAS 4345-03-3
Molecular Formula C33H54O5
Molecular Weight (g/mol) 530.79
MDL Number MFCD00072055
InChI Key IELOKBJPULMYRW-NJQVLOCASA-N
Synonym Vitamin E Succinate
PubChem CID 20353
IUPAC Name 4-oxo-4-[[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl]oxy]butanoic acid
SMILES CC1=C2C(=C(C(=C1C)OC(=O)CCC(=O)O)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C

Specifications

Melting Point 75°C
Color White-Yellow
Quantity 5 g
Formula Weight 530.79
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material D-alpha-Tocopherol Succinate
EINECSNumber (9)-0477
RTECSNumber EJ9984000
TSCA Yes
Recommended Storage Refrigerator