missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ D(-)-alpha-Phenylglycine, 98%
Chemical Identifiers
| CAS | 875-74-1 |
|---|---|
| Molecular Formula | C8H9NO2 |
| Molecular Weight (g/mol) | 151.16 |
| MDL Number | MFCD00008061 |
| InChI Key | ZGUNAGUHMKGQNY-SSDOTTSWSA-N |
| Synonym | d-phenylglycine, d---alpha-phenylglycine, h-d-phg-oh, d-2-phenylglycine, r-2-amino-2-phenylacetic acid, r-phenylglycine, r---2-phenylglycine, d-alpha-phenylglycine, 2r-2-amino-2-phenylacetic acid, 2r-amino phenyl acetic acid |
| PubChem CID | 70134 |
| ChEBI | CHEBI:44962 |
| IUPAC Name | (2R)-2-amino-2-phenylacetic acid |
| SMILES | C1=CC=C(C=C1)C(C(=O)O)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC130630250
|
Thermo Scientific™
130630250 |
25g | Plastic bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC130631000
|
Thermo Scientific™
130631000 |
100g | Plastic bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 875-74-1 | |
| 151.16 | |
| ZGUNAGUHMKGQNY-SSDOTTSWSA-N | |
| 70134 | |
| (2R)-2-amino-2-phenylacetic acid |
| C8H9NO2 | |
| MFCD00008061 | |
| d-phenylglycine, d---alpha-phenylglycine, h-d-phg-oh, d-2-phenylglycine, r-2-amino-2-phenylacetic acid, r-phenylglycine, r---2-phenylglycine, d-alpha-phenylglycine, 2r-2-amino-2-phenylacetic acid, 2r-amino phenyl acetic acid | |
| CHEBI:44962 | |
| C1=CC=C(C=C1)C(C(=O)O)N |
Specifications
| 875-74-1 | |
| Authentic | |
| Plastic bottle | |
| C6H5CH(NH2)CO2H | |
| 25g | |
| 15, 7402 | |
| d-phenylglycine, d---alpha-phenylglycine, h-d-phg-oh, d-2-phenylglycine, r-2-amino-2-phenylacetic acid, r-phenylglycine, r---2-phenylglycine, d-alpha-phenylglycine, 2r-2-amino-2-phenylacetic acid, 2r-amino phenyl acetic acid | |
| ZGUNAGUHMKGQNY-SSDOTTSWSA-N | |
| (2R)-2-amino-2-phenylacetic acid | |
| 70134 | |
| 151.16 | |
| D(-)-α-Phenylglycine, 98% |
| 0.03 max. (c=6, 1M HCl) at 420nm | |
| 98% | |
| C8H9NO2 | |
| MFCD00008061 | |
| 14, 460 | |
| −151° to −161° (20°C, 589nm) (c=5, 1M HCl) | |
| Solubility in water: 0.3g/100mL. Other solubilities: soluble in aq.sol. of inorg. solvents and alkalies,practically insoluble in alcohol | |
| C1=CC=C(C=C1)C(C(=O)O)N | |
| 151.16 | |
| CHEBI:44962 | |
| 98% |
Safety and Handling
EINECSNumber : 212-876-3
TSCA : TSCA
RUO â Research Use Only