missing translation for 'onlineSavingsMsg'
Learn More

D-Allothreonine 99.0+%, TCI America™

Catalog No. A1518100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1518100MG 100 mg
A15181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1518100MG Supplier TCI America Supplier No. A1518100MG

Chemical Identifiers

CAS 24830-94-2
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00004526
InChI Key AYFVYJQAPQTCCC-STHAYSLISA-N
Synonym d-allo-threonine, d-allothreonine, h-d-allo-thr-oh, d--allo-threonine, r-allothreonine, 2r,3r-2-amino-3-hydroxybutanoic acid, 2r,3r-allothreonine, r-allo-threonine, dl-allo-threonine, allothreonine, d
PubChem CID 90624
ChEBI CHEBI:32826
IUPAC Name (2R,3S)-2-amino-3-hydroxybutanoic acid
SMILES C[C@H](O)[C@@H](N)C(O)=O

Specifications

Color White
Quantity 100 mg
Formula Weight 119.12
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material D-Allothreonine
RTECSNumber BA4050000
TSCA No