missing translation for 'onlineSavingsMsg'
Learn More

D-Alloisoleucine 98.0+%, TCI America™

Catalog No. A02121G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A02121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A02121G Supplier TCI America Supplier No. A02121G

Chemical Identifiers

CAS 1509-35-9
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00066445
InChI Key AGPKZVBTJJNPAG-CRCLSJGQSA-N
Synonym d-alloisoleucine, d-allo-isoleucine, 2r,3s-2-amino-3-methylpentanoic acid, allo-d-isoleucine, alloisoleucine, d, h-d-allo-ile-oh, threo-d-isoleucine, dl-allo-isoleucine, hile, h-dl-allo-ile-oh
PubChem CID 94206
ChEBI CHEBI:20899
IUPAC Name (2R,3S)-2-amino-3-methylpentanoic acid
SMILES CC[C@H](C)[C@@H](N)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 131.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Alloisoleucine
EINECSNumber (9)-1560
RTECSNumber BA2930000
TSCA No