missing translation for 'onlineSavingsMsg'
Learn More

D-allo-Threonine, 99%

CAS: 24830-94-2 | C4H9NO3 | 119.12 g/mol

Supplier:  Thermo Scientific Chemicals H2705003

Quantity 1 g

 View more versions of this product

Catalog No. AAH2705003


Only null left

Description

Description

They are found as constituents of an increasing group of biologically active peptides. Stereocontrolled syntheses of chiral and racemic key intermediates to thienamycin from d-allo-threonine and trans-crotonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
They are found as constituents of an increasing group of biologically active peptides. Stereocontrolled syntheses of chiral and racemic key intermediates to thienamycin from d-allo-threonine and trans-crotonic acid.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

24830-94-2
119.12
AYFVYJQAPQTCCC-STHAYSLISA-N
90624
(2R,3R)-2-amino-3-hydroxybutanoic acid
C4H9NO3
MFCD00004526
d-allo-threonine, d-allothreonine, h-d-allo-thr-oh, d--allo-threonine, r-allothreonine, 2r,3r-2-amino-3-hydroxybutanoic acid, 2r,3r-allothreonine, r-allo-threonine, dl-allo-threonine, allothreonine, d
CHEBI:32826
C[C@H](O)[C@@H](N)C(O)=O

Specifications

268°C (decomposition)
24830-94-2
MFCD00004526
d-allo-threonine, d-allothreonine, h-d-allo-thr-oh, d--allo-threonine, r-allothreonine, 2r,3r-2-amino-3-hydroxybutanoic acid, 2r,3r-allothreonine, r-allo-threonine, dl-allo-threonine, allothreonine, d
AYFVYJQAPQTCCC-STHAYSLISA-N
(2R,3R)-2-amino-3-hydroxybutanoic acid
119.12
CHEBI:32826
−8.7° (c=2 in Water)
D-allo-Threonine
C4H9NO3
1721644
Soluble in water.
C[C@H](O)[C@@H](N)C(O)=O
1 g
90624
119.12
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 246-488-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions