missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-allo-Isoleucine, 97%

Catalog No. AC357435000
Change view
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC357435000 500 mg Glass Bottle
AC357430010 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC357435000 Supplier Thermo Scientific Chemicals Supplier No. 357435000

CAS: 1509-35-9 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1509-35-9
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00066445
InChI Key AGPKZVBTJJNPAG-CRCLSJGQSA-N
Synonym d-alloisoleucine, d-allo-isoleucine, 2r,3s-2-amino-3-methylpentanoic acid, allo-d-isoleucine, alloisoleucine, d, h-d-allo-ile-oh, threo-d-isoleucine, dl-allo-isoleucine, hile, h-dl-allo-ile-oh
PubChem CID 94206
ChEBI CHEBI:20899
IUPAC Name (2R,3S)-2-amino-3-methylpentanoic acid
SMILES CC[C@H](C)[C@@H](N)C(O)=O

Specifications

Melting Point 285.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 500 mg
Specific Rotation − 35.00
Formula Weight 131.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material D-allo-Isoleucine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 216-143-9

RUO – Research Use Only