missing translation for 'onlineSavingsMsg'
Learn More

D-allo-Isoleucine, 97%

Catalog No. AAH27189MD
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27189MD 250 mg
AAH2718906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27189MD Supplier Thermo Scientific Chemicals Supplier No. H27189MD

CAS: 1509-35-9 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1509-35-9
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00066445
InChI Key AGPKZVBTJJNPAG-CRCLSJGQSA-N
Synonym d-alloisoleucine, d-allo-isoleucine, 2r,3s-2-amino-3-methylpentanoic acid, allo-d-isoleucine, alloisoleucine, d, h-d-allo-ile-oh, threo-d-isoleucine, dl-allo-isoleucine, hile, h-dl-allo-ile-oh
PubChem CID 94206
ChEBI CHEBI:20899
IUPAC Name (2R,3S)-2-amino-3-methylpentanoic acid
SMILES CC[C@H](C)[C@@H](N)C(O)=O

Specifications

Melting Point ∽285°C (decomposition)
Beilstein 1721794
Quantity 250 mg
Formula Weight 131.18
Optical Rotation −37° (c=1 in 5N HCl)
Percent Purity 97%
Chemical Name or Material D-allo-Isoleucine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-143-9
RTECSNumber BA2930000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only