missing translation for 'onlineSavingsMsg'
Learn More

D-Alanine Methyl Ester Hydrochloride 97.0+%, TCI America™

Catalog No. A20115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20115G 5 g
A201125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20115G Supplier TCI America Supplier No. A20115G
Only null left

Chemical Identifiers

CAS 14316-06-4
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00066141
InChI Key IYUKFAFDFHZKPI-UHFFFAOYNA-N
Synonym d-alanine methyl ester hydrochloride, methyl d-alaninate hydrochloride, h-d-ala-ome.hcl, methyl d-alaninate hcl, methyl 2r-2-aminopropanoate hydrochloride, h-d-ala-ome hcl, r-methyl 2-aminopropanoate hydrochloride, d-alanine methylester hydrochloride, d-alanine, methyl ester, hydrochloride, pubchem10885
PubChem CID 11182647
IUPAC Name hydrogen methyl 2-aminopropanoate chloride
SMILES [H+].[Cl-].COC(=O)C(C)N

Specifications

Melting Point 108°C
Color White
Quantity 5 g
Formula Weight 139.58
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material D-Alanine Methyl Ester Hydrochloride