missing translation for 'onlineSavingsMsg'
Learn More

D-Alanine methyl ester hydrochloride, 98%

Catalog No. AAH6247222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6247222 100 g
AAH6247206 5 g
AAH6247214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6247222 Supplier Thermo Scientific Chemicals Supplier No. H6247222

CAS: 14316-06-4 | C4H10ClNO2 | 139.58 g/mol

D-Alanine methyl ester hydrochloride is used as building block for the preparation of peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Alanine methyl ester hydrochloride is used as building block for the preparation of peptides.

Solubility
Soluble in Water 100 mg/ml.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 14316-06-4
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00066141
InChI Key IYUKFAFDFHZKPI-UHFFFAOYNA-N
Synonym d-alanine methyl ester hydrochloride, methyl d-alaninate hydrochloride, h-d-ala-ome.hcl, methyl d-alaninate hcl, methyl 2r-2-aminopropanoate hydrochloride, h-d-ala-ome hcl, r-methyl 2-aminopropanoate hydrochloride, d-alanine methylester hydrochloride, d-alanine, methyl ester, hydrochloride, pubchem10885
PubChem CID 11182647
IUPAC Name methyl (2R)-2-aminopropanoate;hydrochloride
SMILES [H+].[Cl-].COC(=O)C(C)N

Specifications

Melting Point 108°C to 110°C
Color White
Beilstein 4235975
Solubility Information Soluble in Water 100mg/ml.
Quantity 100 g
Formula Weight 139.58
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material D-Alanine methyl ester hydrochloride
EINECSNumber 238-258-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only