missing translation for 'onlineSavingsMsg'
Learn More

D-Alanine Benzyl Ester p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. a20045g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20045G 5 g
A200425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20045G Supplier TCI America Supplier No. A20045G

Chemical Identifiers

CAS 41036-32-2
Molecular Formula C17H21NO5S
Molecular Weight (g/mol) 351.42
MDL Number MFCD00039092
InChI Key NWOPHJSSBMABBD-DDWIOCJRSA-N
Synonym h-d-ala-obzl.tosoh, d-alanine benzyl ester p-toluenesulfonate salt, d-alanine benzyl ester p-toluenesulfonate, d-alanine benzyl ester tosylate, h-d-ala-obzl p-tosylate, h-d-ala-obzl.tos-oh, o-benzyl-d-alanine toluene-p-sulphonate, r-benzyl 2-aminopropanoate 4-methylbenzenesulfonate, benzyl 2r-2-aminopropanoate; para-toluene sulfonate, d-ala-obzltosoh
PubChem CID 2802425
IUPAC Name 4-methylbenzene-1-sulfonic acid; benzyl (2R)-2-aminopropanoate
SMILES CC1=CC=C(C=C1)S(O)(=O)=O.C[C@@H](N)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 5 g
Formula Weight 351.42
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material D-Alanine Benzyl Ester p-Toluenesulfonate