missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Alanine, 99+%

CAS: 338-69-2 | C3H7NO2 | 89.09 g/mol

Supplier:  Thermo Scientific Chemicals 172080250

Quantity 25 g
Packaging Glass Bottle

 View more versions of this product

Catalog No. AC172080250


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

338-69-2
89.09
QNAYBMKLOCPYGJ-UWTATZPHSA-N
71080
(2R)-2-aminopropanoic acid
C3H7NO2
MFCD00008077
d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form
CHEBI:15570
C[C@@H](N)C(O)=O

Specifications

D-Alanine
338-69-2
White
10ppm max.
0.2% max. (105°C, 3 hrs)
Glass Bottle
CH3CH(NH2)CO2H
− 14.50
d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form
Solubility in water: 155g/L in water (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and organic solvents
C[C@@H](N)C(O)=O
89.09
71080
89.09
Crystalline Powder or Crystals
278.0°C to 282.0°C
2ppm max. (As2O3)
0.1% max.
Authentic
99+%
C3H7NO2
MFCD00008077
95% min. (c=10, H2O, 430nm) 1cm cell
0.2% max.
QNAYBMKLOCPYGJ-UWTATZPHSA-N
(2R)-2-aminopropanoic acid
25 g
CHEBI:15570
≥99%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions