missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-3-Phenyllactic Acid 98.0+%, TCI America™
$76.43 - $233.00
Chemical Identifiers
| CAS | 7326-19-4 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.18 |
| MDL Number | MFCD00078062 |
| InChI Key | VOXXWSYKYCBWHO-MRVPVSSYSA-N |
| Synonym | r-2-hydroxy-3-phenylpropionic acid, r-2-hydroxy-3-phenylpropanoic acid, r-2-hydroxy-3-phenylpropanoic acid, d-+-phenyllactic acid, d-+-3-phenyllactic acid, d-+-3-phenyllactic acid, r-phenyllactate, 3-phenyl-d-lactic acid, 3-phenyl-d-lactic acid, r-, r- |
| PubChem CID | 643327 |
| ChEBI | CHEBI:32978 |
| IUPAC Name | (2R)-2-hydroxy-3-phenylpropanoic acid |
| SMILES | O[C@H](CC1=CC=CC=C1)C(O)=O |
Chemical Identifiers
| 7326-19-4 | |
| 166.18 | |
| VOXXWSYKYCBWHO-MRVPVSSYSA-N | |
| 643327 | |
| (2R)-2-hydroxy-3-phenylpropanoic acid |
| C9H10O3 | |
| MFCD00078062 | |
| r-2-hydroxy-3-phenylpropionic acid, r-2-hydroxy-3-phenylpropanoic acid, r-2-hydroxy-3-phenylpropanoic acid, d-+-phenyllactic acid, d-+-3-phenyllactic acid, d-+-3-phenyllactic acid, r-phenyllactate, 3-phenyl-d-lactic acid, 3-phenyl-d-lactic acid, r-, r- | |
| CHEBI:32978 | |
| O[C@H](CC1=CC=CC=C1)C(O)=O |
Specifications
| 7326-19-4 | |
| White-Yellow | |
| MFCD00078062 | |
| r-2-hydroxy-3-phenylpropionic acid, a-hydroxy-3-phenylpropionic acid, r-2-hydroxy-3-phenylpropanoic acid, d-+-phenyllactic acid, d-+-phenyllactic acid, d-+-3-phenyllactic acid, r-+-3-phenyllactic acid, r-+-3-phenyllactic acid, r-3-phenyllactic acid, r-, r- | |
| O[C@H](CC1=CC=CC=C1)C(O)=O | |
| 166.18 | |
| CHEBI:32978 | |
| ≥98.0% (HPLC,T) | |
| D-(+)-3-Phenyllactic Acid |
| 125°C | |
| C9H10O3 | |
| 1 g | |
| VOXXWSYKYCBWHO-MRVPVSSYSA-N | |
| (2R)-2-hydroxy-3-phenylpropanoic acid | |
| 643327 | |
| 166.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No