missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-3-Phenyllactic Acid 98.0+%, TCI America™

Catalog No. P19811G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19811G 1 g
P19815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P19811G Supplier TCI America Supplier No. P19811G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7326-19-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00078062
InChI Key VOXXWSYKYCBWHO-MRVPVSSYSA-N
Synonym r-2-hydroxy-3-phenylpropionic acid, r-2-hydroxy-3-phenylpropanoic acid, 2r-2-hydroxy-3-phenylpropanoic acid, d-+-phenyllactic acid, d-+-3-phenyllactic acid, r-phenyllactate, r-+-3-phenyllactic acid, 3-phenyl-d-lactic acid, r-3-phenyllactic acid, r-, a-hydroxy-3-phenylpropionic acid
PubChem CID 643327
ChEBI CHEBI:32978
IUPAC Name (2R)-2-hydroxy-3-phenylpropanoic acid
SMILES O[C@H](CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 1 g
Formula Weight 166.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-3-Phenyllactic Acid