missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-2-Phenylglycine 99.0+%, TCI America™
$38.51 - $267.61
Chemical Identifiers
| CAS | 875-74-1 |
|---|---|
| Molecular Formula | C8H9NO2 |
| Molecular Weight (g/mol) | 151.165 |
| MDL Number | MFCD00008061 |
| InChI Key | ZGUNAGUHMKGQNY-SSDOTTSWSA-N |
| Synonym | d-phenylglycine, d---alpha-phenylglycine, h-d-phg-oh, d-2-phenylglycine, r-2-amino-2-phenylacetic acid, r-phenylglycine, r---2-phenylglycine, d-alpha-phenylglycine, 2r-2-amino-2-phenylacetic acid, 2r-amino phenyl acetic acid |
| PubChem CID | 70134 |
| ChEBI | CHEBI:44962 |
| IUPAC Name | (2R)-2-amino-2-phenylacetic acid |
| SMILES | C1=CC=C(C=C1)C(C(=O)O)N |
Chemical Identifiers
| 875-74-1 | |
| 151.165 | |
| ZGUNAGUHMKGQNY-SSDOTTSWSA-N | |
| 70134 | |
| (2R)-2-amino-2-phenylacetic acid |
| C8H9NO2 | |
| MFCD00008061 | |
| d-phenylglycine, d---alpha-phenylglycine, h-d-phg-oh, d-2-phenylglycine, r-2-amino-2-phenylacetic acid, r-phenylglycine, r---2-phenylglycine, d-alpha-phenylglycine, 2r-2-amino-2-phenylacetic acid, 2r-amino phenyl acetic acid | |
| CHEBI:44962 | |
| C1=CC=C(C=C1)C(C(=O)O)N |
Specifications
| 875-74-1 | |
| C8H9NO2 | |
| 25 g | |
| ZGUNAGUHMKGQNY-SSDOTTSWSA-N | |
| (2R)-2-amino-2-phenylacetic acid | |
| 70134 | |
| 151.17 | |
| Crystalline Powder |
| White | |
| MFCD00008061 | |
| d-phenylglycine, d---alpha-phenylglycine, h-d-phg-oh, d-2-phenylglycine, r-2-amino-2-phenylacetic acid, r-phenylglycine, r---2-phenylglycine, d-alpha-phenylglycine, 2r-2-amino-2-phenylacetic acid, 2r-amino phenyl acetic acid | |
| C1=CC=C(C=C1)C(C(=O)O)N | |
| 151.165 | |
| CHEBI:44962 | |
| ≥99.0% (T) | |
| D-2-Phenylglycine |
Safety and Handling
EINECSNumber : (3)-1441
TSCA : Yes