missing translation for 'onlineSavingsMsg'
Learn More

D-2-Phenylglycine 99.0+%, TCI America™

Catalog No. P082025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P082025G 25 g
P0820100G 100 g
P0820500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P082025G Supplier TCI America Supplier No. P082025G
Only null left

Chemical Identifiers

CAS 875-74-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00008061
InChI Key ZGUNAGUHMKGQNY-SSDOTTSWSA-N
Synonym d-phenylglycine, d---alpha-phenylglycine, h-d-phg-oh, d-2-phenylglycine, r-2-amino-2-phenylacetic acid, r-phenylglycine, r---2-phenylglycine, d-alpha-phenylglycine, 2r-2-amino-2-phenylacetic acid, 2r-amino phenyl acetic acid
PubChem CID 70134
ChEBI CHEBI:44962
IUPAC Name (2R)-2-amino-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)N

Specifications

Color White
Quantity 25 g
Formula Weight 151.17
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-2-Phenylglycine
EINECSNumber (3)-1441
TSCA Yes