missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-2-Phenylglycine Methyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. P259225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P259225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P259225G Supplier TCI America Supplier No. P259225G

Chemical Identifiers

CAS 19883-41-1
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD00137487
InChI Key DTHMTBUWTGVEFG-VGVXOIKUNA-N
Synonym h-d-phg-ome.hcl, h-d-phg-ome hcl, r-methyl 2-amino-2-phenylacetate hydrochloride, r-2-phenylglycine methyl ester hydrochloride, r---2-phenylglycine methyl ester hydrochloride, methyl 2r-amino phenyl acetate hydrochloride, d-alpha-phenylglycine methyl ester hydrochloride, d---2-phenylglycine methyl ester hydrochloride, methyl 2r-2-amino-2-phenylacetate hydrochloride, d-phenylglycine methyl ester hydrochloride
PubChem CID 12247453
IUPAC Name methyl (2R)-2-amino-2-phenylacetate hydrochloride
SMILES Cl.COC(=O)[C@H](N)C1=CC=CC=C1

Specifications

Melting Point 219°C
Color White-Yellow
Quantity 25 g
Formula Weight 201.65
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material D-(-)-2-Phenylglycine Methyl Ester Hydrochloride