missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-2-Phenylglycine methyl ester hydrochloride, 96%

Catalog No. AAL1873514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1873514 25 g
AAL1873522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1873514 Supplier Thermo Scientific Chemicals Supplier No. L1873514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19883-41-1 | C9H12ClNO2 | 201.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19883-41-1
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD00137487
InChI Key DTHMTBUWTGVEFG-VGVXOIKUNA-N
Synonym h-d-phg-ome.hcl, h-d-phg-ome hcl, r-methyl 2-amino-2-phenylacetate hydrochloride, r-2-phenylglycine methyl ester hydrochloride, r---2-phenylglycine methyl ester hydrochloride, methyl 2r-amino phenyl acetate hydrochloride, d-alpha-phenylglycine methyl ester hydrochloride, d---2-phenylglycine methyl ester hydrochloride, methyl 2r-2-amino-2-phenylacetate hydrochloride, d-phenylglycine methyl ester hydrochloride
PubChem CID 12247453
IUPAC Name methyl (2R)-2-amino-2-phenylacetate hydrochloride
SMILES Cl.COC(=O)[C@H](N)C1=CC=CC=C1

Specifications

Melting Point ∽190°C (decomposition)
Beilstein 3915654
Quantity 25 g
Formula Weight 201.65
Optical Rotation −118° (c=1 in Water)
Percent Purity 96%
Chemical Name or Material D-(-)-2-Phenylglycine methyl ester hydrochloride
EINECSNumber 243-399-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only