missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-(-)-2-Phenylglycine, 99%
CAS: 875-74-1 | C8H9NO2 | 151.165 g/mol
$51.19 - $517.23
Chemical Identifiers
| CAS | 875-74-1 |
|---|---|
| Molecular Formula | C8H9NO2 |
| Molecular Weight (g/mol) | 151.165 |
| MDL Number | MFCD00008061 |
| InChI Key | ZGUNAGUHMKGQNY-SSDOTTSWSA-N |
| Synonym | d-phenylglycine, d---alpha-phenylglycine, h-d-phg-oh, d-2-phenylglycine, r-2-amino-2-phenylacetic acid, r-phenylglycine, r---2-phenylglycine, d-alpha-phenylglycine, 2r-2-amino-2-phenylacetic acid, 2r-amino phenyl acetic acid |
| PubChem CID | 70134 |
| ChEBI | CHEBI:44962 |
| IUPAC Name | (2R)-2-amino-2-phenylacetic acid |
| SMILES | C1=CC=C(C=C1)C(C(=O)O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1566914
|
Thermo Scientific Chemicals
A1566914 |
25 g |
Each for $51.19
|
|
|||||
|
AAA1566922
|
Thermo Scientific Chemicals
A1566922 |
100 g |
Each for $137.70
|
|
|||||
|
AAA1566936
|
Thermo Scientific Chemicals
A1566936 |
500 g |
Each for $517.23
|
|
|||||
Description
D-(-)-2-Phenylglycine is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors. Used to prepare semisynthetic antibiotics amoxicillin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-(-)-2-Phenylglycine is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors. Used to prepare semisynthetic antibiotics amoxicillin.
Solubility
Soluble in water (0.3g/100mL) and aqueous acid.
Notes
Store at room temperature. Store in cool dry place in tightly closed container. Store away from oxidizing agent.
Chemical Identifiers
| 875-74-1 | |
| 151.165 | |
| ZGUNAGUHMKGQNY-SSDOTTSWSA-N | |
| 70134 | |
| (2R)-2-amino-2-phenylacetic acid |
| C8H9NO2 | |
| MFCD00008061 | |
| d-phenylglycine, d---alpha-phenylglycine, h-d-phg-oh, d-2-phenylglycine, r-2-amino-2-phenylacetic acid, r-phenylglycine, r---2-phenylglycine, d-alpha-phenylglycine, 2r-2-amino-2-phenylacetic acid, 2r-amino phenyl acetic acid | |
| CHEBI:44962 | |
| C1=CC=C(C=C1)C(C(=O)O)N |
Specifications
| 277°C (decomposition) | |
| C8H9NO2 | |
| 2208676 | |
| d-phenylglycine, d---alpha-phenylglycine, h-d-phg-oh, d-2-phenylglycine, r-2-amino-2-phenylacetic acid, r-phenylglycine, r---2-phenylglycine, d-alpha-phenylglycine, 2r-2-amino-2-phenylacetic acid, 2r-amino phenyl acetic acid | |
| ZGUNAGUHMKGQNY-SSDOTTSWSA-N | |
| (2R)-2-amino-2-phenylacetic acid | |
| 151.165 | |
| CHEBI:44962 | |
| 151.17 | |
| D-(-)-2-Phenylglycine |
| 875-74-1 | |
| MFCD00008061 | |
| 14,7291 | |
| Soluble in water (0.3g/100mL) and aqueous acid. | |
| C1=CC=C(C=C1)C(C(=O)O)N | |
| 25 g | |
| 70134 | |
| −154° (c=1 in 1N HCl) | |
| 99% |
Safety and Handling
EINECSNumber : 212-876-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only