missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(-)-2-Phenylglycine, 99%

Catalog No. AAA1566936
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1566936 500 g
AAA1566914 25 g
AAA1566922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1566936 Supplier Thermo Scientific Chemicals Supplier No. A1566936
Only null left

CAS: 875-74-1 | C8H9NO2 | 151.165 g/mol

D-(-)-2-Phenylglycine is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors. Used to prepare semisynthetic antibiotics amoxicillin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-2-Phenylglycine is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors. Used to prepare semisynthetic antibiotics amoxicillin.

Solubility
Soluble in water (0.3g/100mL) and aqueous acid.

Notes
Store at room temperature. Store in cool dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 875-74-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00008061
InChI Key ZGUNAGUHMKGQNY-SSDOTTSWSA-N
Synonym d-phenylglycine, d---alpha-phenylglycine, h-d-phg-oh, d-2-phenylglycine, r-2-amino-2-phenylacetic acid, r-phenylglycine, r---2-phenylglycine, d-alpha-phenylglycine, 2r-2-amino-2-phenylacetic acid, 2r-amino phenyl acetic acid
PubChem CID 70134
ChEBI CHEBI:44962
IUPAC Name (2R)-2-amino-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)N

Specifications

Melting Point 277°C (decomposition)
Beilstein 2208676
Merck Index 14,7291
Solubility Information Soluble in water (0.3g/100mL) and aqueous acid.
Quantity 500 g
Formula Weight 151.17
Optical Rotation −154° (c=1 in 1N HCl)
Percent Purity 99%
Chemical Name or Material D-(-)-2-Phenylglycine
EINECSNumber 212-876-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only