missing translation for 'onlineSavingsMsg'
Learn More

D-2-Cyclohexylglycine 98.0+%, TCI America™

Catalog No. C26731G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C26731G Supplier TCI America Supplier No. C26731G
Only null left

Chemical Identifiers

CAS 14328-52-0
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD01311678
InChI Key WAMWSIDTKSNDCU-SSDOTTSWSA-N
Synonym d-cyclohexylglycine, d-alpha-cyclohexylglycine, r-2-amino-2-cyclohexylacetic acid, h-d-chg-oh, 2-cyclohexyl-d-glycine, 2r-amino cyclohexyl acetic acid, 2r-2-amino-2-cyclohexylacetic acid, d-2-cyclohexylglycine, d-cyclohexyl glycine, h-cyclohexyl-d-gly-oh
PubChem CID 736849
IUPAC Name (2R)-2-amino-2-cyclohexylacetic acid
SMILES C1CCC(CC1)C(C(=O)O)N

Specifications

Melting Point 253°C
Color White
Quantity 1 g
Formula Weight 157.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material D-2-Cyclohexylglycine